Alfa Aesar™ 3-Trimethylsilylpropynal, 97%

Manufacturer: Alfa Aesar™ 4476301

Quantity 0.25g

 View more versions of this product

Catalog No. AA4476301

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2975-46-4
    Molecular Formula C6H10OSi
    Linear Formula (CH3)3SiC≡CCHO
    Formula Weight 126.23
    MDL Number MFCD01103709
    UN Number UN1993
    Chemical Name or Material 3-Trimethylsilylpropynal

    Specifications

    Formula Weight 126.23
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    Molecular Formula C6H10OSi
    Refractive Index 1.4435
    Linear Formula (CH3)3SiC≡CCHO
    MDL Number MFCD01103709
    Color Colorless to yellow
    Quantity 0.25g
    CAS 2975-46-4
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Recommended Storage 2°C to 8°C
    UN Number UN1993
    Storage Note 1
    • Packaged under Argon
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Density 0.86g/mL
    Sensitivity Air & Moisture Sensitive
    Boiling Point 58° to 60°C (37.5mmHg)
    Health Hazard 1 Warning
    Flash Point 29°C (84°F)
    Chemical Name or Material 3-Trimethylsilylpropynal
    Assay 97%

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2°C to 8°C

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit