Learn More
Medchemexpress LLC 3-(Tritylthio)propionic acid | 27144-18-9 | MFCD00237291 | 99.77% | 348.46 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW011156
Encompass_Preferred
Mpa(Trt) is a 3-mercaptopropionic acid derivative containing a trityl protecting group (Trt) and can be used to synthesize compounds with anti-leukemia activity.
  • Used to synthesize compounds with anti-leukemia activity.
  • High purity: 99.77%.
  • Available in various sizes: 25 g, 100 g, 500 g.
  • Research use only.
Catalog No. 50-003-66889