Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Undecyne, 97%

Click to view available options
Quantity:
5g
25g
60212-30-8
C11H20
152.281
MFCD00041663
DPWGJNPCPLQVKQ-UHFFFAOYSA-N
3-undecyne,acmc-209mii,dpwgjnpcplqvkq-uhfffaoysa
143689
undec-3-yne
CCCCCCCC#CCC
60212-30-8
C11H20
152.281
MFCD00041663
DPWGJNPCPLQVKQ-UHFFFAOYSA-N
3-undecyne,acmc-209mii,dpwgjnpcplqvkq-uhfffaoysa
143689
undec-3-yne
CCCCCCCC#CCC
Boiling Point 79°C to 81°C (10mmHg)
Quantity 5g
Formula Weight 152.28
Percent Purity 97%
Chemical Name or Material 3-Undecyne

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning