Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ergodyne™ ProFlex™ 925CR6 Performance Dorsal Impact-Reducing and Cut Resistant Gloves
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
- Next-gen Armortex™-backed palm for extreme cut protection
- Full TPR armor and technical foam for max protection
- Hi-Vis AX Suede™ and non-slip dot gripping palm
- Armortex™-reinforced thumb saddle and index finger
- Padded neoprene cuff with reflective binding
- Pull-on tab with ID space
Specifications
Color | Lime Green |
Cut Resistance Rating | ANSI A6 |
Material | Next-gen Armortex |
Product Type | Cut-resistant Gloves |
Size | X-Large |
EN388 Standard | EN 388:4443 |
For Use With (Application) | Oil/Gas Drilling, Mining, Heavy Construction, Ironwork |
Glove Cuff Color | Black, Gray |
Glove Cuff Style | Padded neoprene cuff with reflective binding |
Liner Material | TPR Armor, Technical Foam |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction