Learn More
Justrite™ 30 Gallon ChemCor™ Corrosives/Acids Safety Cabinet With Bi-Fold Self-Closing Doors
GSA_VA
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 30 Gallon ChemCor™ Corrosives/Acids Safety Cabinet With Bi-Fold Self-Closing Doors offers superior corrosion protection combined with fire protection in a single cabinet.
$5182.48 - $6589.28
Specifications
Color Blue
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Price Quantity & Availability  
19-822-317
GSA_VA
View Documents
Justrite™
8930822
30 gal. 114 L
Each for $5,182.48
Add to cart
19-822-318
GSA_VA
View Documents
Justrite™
8945822
45 gal. 170 L
Each for $6,589.28
Add to cart
Description
Description
  • Lid opens to no more than 60 degrees and stays closed when not in use, isolating contents from fire sources and limiting oxygen to virtually eliminate the risk of spontaneous combustion
  • Round construction and elevated base allow circulation of air around can to disperse heat and Reduce moisture build-up and rusting
  • Durable steel construction with a powder-coat finish provides chemical resistance
  • Approved by FM, UL, and TUV
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Blue