Learn More
Bullard™ 3000 Series Patriot Safety Helmet
SDP
Supplier:  Bullard™ TT2
 View more versions of this product
Catalog No. 19-084463
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
The 3000 Series Patriot Safety Helmet is ideal for almost any industrial application, and its American Flag design will let you proudly demonstrate your patriotism. Permanent full graphic coverage will not fade, peel, or crack. Choose the six-point ratchet or pinlock suspension to provide a comfortable and secure fit. Accessory slots accept faceshields, hearing protection, welding helmets, or communication devices. Replaceable Sportek™ wicking browpad comes standard. Also available: replacement ratchet and pinlock suspensions, and replacement browpads.
Compliance
Meets ANSI Z89.1-1997 Type I, Class E, G and C requirements.
Specifications
Specifications
Meets ANSI Z89.1-1997 Type I; Class E, G and C.
6-Point Ratchet or Pin-Lock
Product Suggestions
Product Suggestions