Learn More
Thermo Scientific™ 316 Stainless Steel Capillary Tubing for HPLC Available on GSA/VA Contract for Federal Government customers only.
Use cleaned, polished, passivated and ready-to-use Thermo Scientific? 316 Stainless Steel Capillary Tubing for HPLC.
Manufacturer:  Thermo Scientific™ U155
 View more versions of this product
Catalog No. 03-170-430
  Check Availability
Add to cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Use Thermo Scientific? 316 Stainless Steel Capillary Tubing for ultra-high pressure applications. Prefinished, square, burr-free ends and interiors ensure minimum dead volume connections. Cleaned, polished and passivated, it is ready for immediate use. This tubing is not recommended for biological samples.
  • Suitable for ultra high pressure applications
  • Wide chemical compatibilty
  • Prefinished, square, burr-free ends and interiors to minimize dead volume connections
Description & Specifications
Specifications
0.005 in. ID; 30cm length; Red
HPLC
0.005 in. ID; 30cm length; Red
HPLC
Capillary Tubing