Thermo Scientific™ 316 Stainless Steel Capillary Tubing for HPLC in 5-Foot Coils Available on GSA/VA Contract for Federal Government customers only.
Use cleaned, polished and passivated Thermo Scientific™ 316 Stainless Steel Capillary Tubing for HPLC, ready for immediate use.
Manufacturer: Thermo Scientific™ U108
 View more versions of this product
Catalog No. 03-170-404
  Check Availability
Add to Cart
Alert
  • Not recommended for biological samples
  • Rough internal surface may lead to sample carryover
  • Reasonably flexible, but works best with minimal bending
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    For Use With HPLC
    Type Capillary Tubing
    Item Description 0.007 in. ID; 5 ft. length
    Description 0.007 in. ID; 5 ft. length
    Use Thermo Scientific™ 316 Stainless Steel Capillary Tubing for ultrahigh-pressure applications. Prefinished, square, burr-free ends and interiors ensure minimum dead volume connections. Cleaned, polished and passivated, it is ready for immediate use. This tubing is not recommended for biological samples.
    • Suitable for ultra high pressure applications
    • Wide chemical compatibilty
  • Description & Specifications
Related Products
Product Content Feedback