Learn More
CEM Corporation 35ML SP-D PLUS ATTENUATOR
SDP
Manufacturer:  CEM Corporation 526370
Encompass
35 mL Discover SP-D Plus AttenuatorAnalytical Teflon coated corrosion resistant attenuator for use with the 35 mL reaction vial.
Catalog No. NC1457977