Thermo Scientific™ White and Black 384-Well Immuno Plates Available on GSA/VA Contract for Federal Government customers only.

Conduct quantitative and qualitative immunoassays with Thermo Scientific™ White and Black 384-Well Immuno Plates.

Manufacturer: Thermo Scientific™ 460372

 View more versions of this product

Catalog No. 12-565-345

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Binding Property Adsorption,Hydrophilic
    Surface Coating MaxiSorp,Polystyrene
    Plate Blocking None
    Detection Method Chemiluminescent
    Format 384-well plate
    For Use With (Application) Fluorescence and Luminescence
    No. per Case 30
    No. per Pack 10
    Volume (Metric) Well 120µL
    Volume (Metric) Working 100µL
    Well Design Flat
    Color White
    Applications Fluorescence and Luminescence
    Well Shape Flat

    Conduct quantitative and qualitative immunoassays with Thermo Scientific™ White and Black 384-Well Immuno Plates. Ideal for ELISA and binding assays, these plates are compatible with common instruments. They offer lot-to-lot reliability and well-to-well reproducibility for optimal results.

    • Plate has SBS footprint — fits standard equipment
    • Available with flat-bottom wells
    • MaxiSorp™ is hydrophilic and ideal for antibody sandwich assays
    • Offered as black or white plates for optimal choice of application type
    • Certified binding homogeneity/reproducibility

    White

    • Gives maximum reflection
    • Minimum autofluorescence and autoluminescence

    Black

    • Minimizes background in fluorescence reading
    • Minimizes back-scattered light
    • Minimizes crosstalk

    Compatible with:

    Compatible with lids for MicroWell™ plates, sealing tape and breathable membrane

    Ordering Information:

    Please note availability may vary by country

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit