Learn More
Corning™ 384-Well Low Flange Polystyrene Microplates
For use with fluorescent assays with detection microplate reader. Corning™ 384-Well Low Flange Polystyrene Microplates have low background fluorescence, minimal light scatter and reduced crosstalk.
$3710.00
Specifications
Format Standard
Dimensions (L x W x H) 127.8 x 85.5 x 14.35 mm (5 x 3.36 x 0.56 in.)
Bottom Flat
View More Specs
Products 1
Catalog Number Mfr. No. Surface Treatment Sterility Color Price Quantity & Availability  
Catalog Number Mfr. No. Surface Treatment Sterility Color Price Quantity & Availability  
07-201-445 View Documents Corning™
3769
Non-binding surface (NBS) Non-sterile White Case of 100 for $3,710.00
Add to cart
Description
Description
  • Optically clear, flat well bottom permits direct microscope viewing
  • Used with both top and bottom reading instruments
  • TCT treated for optimal cell attachment
  • Individual alphanumeric codes for well identification
  • Opaque walls to prevent well-to-well crosstalk
  • Low flange base designed for barcode customization and robotic handling
  • 07-201-446, -447, -448, -449 have black walls which allow low background fluorescence and minimal light scatter
Specifications
Specifications
Standard
127.8 x 85.5 x 14.35 mm (5 x 3.36 x 0.56 in.)
Flat
Safety and Handling
Safety and Handling
ShelfLife : 2000 hrs.