Learn More
Thermo Scientific™ 96-Well Plate Plastic Non-Coated, Mid Height, Round Well
For any standard application or assay, you can be sure of excellent solvent resistance and low background noise with these high quality polypropylene 96 well microplates.
Supplier: Thermo Scientific™ 60180P208
Description
Whatever your standard or routine application or assay, you can be sure of excellent solvent resistance and low background noise with Thermo Scientific™ WebSeal 96-Well Microplates. Made of high-quality, GC-tested polypropylene, these microplates are ideal for:
- Pharmaceutical and industrial QA/QC
- High-throughput screening (HTS)
- Sample collection and storage
- Combinatorial chemistry
- Liquid phase assays
Chemically and Thermally Resistant Polypropylene
- Broad chemical compatibility with alcohols, acetonitrile and other common HPLC solvents
- Will tolerate temperatures from −80° C to +121°C
- GC-tested to ensure low extractables
Broad Instrument Compatibility
- SBS and ANSI standard footprint design
C17
- Shallow well and deep well formats
- U- and V- bottom wells ensure optimal sample recovery
Specifications
U | |
8 x 12 | |
Plastic | |
Non-sterile | |
96/Pk. |
Standard | |
Webseal™ | |
96 | |
Non-coated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.