Learn More
Thermo Scientific™ WebSeal 384-Well Non-Coated Plastic Microplates
For any standard application or assay, you can be sure of excellent solvent resistance and low background noise with these high quality polypropylene 384 well microplates.
$98.70 - $1423.00
Specifications
Format Standard
Array 16 x 24
Product Type Plastic, Non-Coated, Non-Sterile Well Plate
Material Polypropylene
No. of Wells 384
View More Specs
Products 10
Catalog Number Mfr. No. Total Volume (Metric) Well Bottom No. per Pack Price Quantity & Availability  
Catalog Number Mfr. No. Total Volume (Metric) Well Bottom No. per Pack Price Quantity & Availability  
03-252-412
View Documents
Thermo Scientific™
60180P107
252 μL U 5
Pack of 5 for $98.70
Add to cart
03-252-416
View Documents
Thermo Scientific™
60180P109
145 μL V 10
Pack of 10 for $147.00
Add to cart
03-252-157
View Documents
Thermo Scientific™
60180P203
58 μL U 60
Case of 60 for $1,040.00
Add to cart
03-252-159
View Documents
Thermo Scientific™
60180P204
300 μL V 48
Case of 48 for $1,423.00
Add to cart
03-252-158
View Documents
Thermo Scientific™
60180P213
58 μL U 10
Pack of 10 for $241.50
Add to cart
03-252-160
View Documents
Thermo Scientific™
60180P214
300 μL V 6
Pack of 6 for $225.00
Add to cart
03-252-410
View Documents
Thermo Scientific™
60180P106
58 μL U 25 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
03-252-411
View Documents
Thermo Scientific™
60180P136
58 μL U 100 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
03-252-415
View Documents
Thermo Scientific™
60180P138
120 μL U 120 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
03-252-413
View Documents
Thermo Scientific™
60180P137
252 μL U 60 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Whatever your standard or routine application or assay, you can be sure of excellent solvent resistance and low background noise with Thermo Scientific™ WebSeal 384-Well Non-Coated Plastic Microplates. Made of high-quality, GC-tested polypropylene.
Ideal for:
  • Pharmaceutical and industrial QA/QC
  • High-throughput screening (HTS)
  • Sample collection and storage
  • Combinatorial chemistry
  • Liquid phase assays
Chemically and Thermally Resistant Polypropylene
  • Broad chemical compatibility with alcohols, acetonitrile and other common HPLC solvents
  • Will tolerate temperatures from −80° C to +121°C
  • GC-tested to ensure low extractables
Broad Instrument Compatibility
  • SBS and ANSI standard footprint design
C17
  • Shallow well and deep well formats
  • U- and V- bottom wells ensure optimal sample recovery
Specifications
Specifications
Standard
Plastic, Non-Coated, Non-Sterile Well Plate
384
Non-coated
16 x 24
Polypropylene
Non-sterile
SDS
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.