Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Greiner Bio-One 384-Well Polystyrene Streptavidin-coated Flat Bottom Microplate
Pre-blocked and ready-to-use.
Specifications
Temperature Range (Metric) Room Temperature
Max. RCF 4800 x g (swinging-bucket rotor)
Sterility Non-sterile
Bottom Flat
Well Shape Rounded Square
View More Specs
Products 4
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
07-000-902
View Documents
Greiner Bio-One
781990
Clear
Case of 40 for $5,060.00
N/A
07-000-903
View Documents
Greiner Bio-One
781995
White
Case of 40 for $4,150.00
N/A
07-000-904
View Documents
Greiner Bio-One
781996
Black, Clear Bottom
Case of 40 for $4,920.00
N/A
07-000-905
View Documents
Greiner Bio-One
781997
Black
Case of 40 for $4,500.00
N/A
Description
Description
  • Manufactured under DIN ISO 9001 guidelines
  • Biotin binding capacity: >1.5ng/well or >6pmol/well
  • Solid bottom
  • Storable at room temperature
  • With lid
Specifications
Specifications
Room Temperature
Non-sterile
Rounded Square
Standard
Not Autoclavable
4800 x g (swinging-bucket rotor)
Flat
Polystyrene
5
Safety and Handling
Safety and Handling
ShelfLife : 3 Years after month of production