Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3A-Amino-3A-deoxy-(2AS,3AS)-beta-cyclodextrin Hydrate 97.0+%, TCI America™

Click to view available options
Quantity:
200mg
1g
117194-77-1
C42H71NO34
1134.003
OZLVLKFBPJMRCM-FUFZOPRGSA-N
11320754
C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(O2)C(C8O)O)CO)CO)CO)CO)CO)CO)O)N)O
117194-77-1
C42H71NO34
1134.003
OZLVLKFBPJMRCM-FUFZOPRGSA-N
11320754
C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(O2)C(C8O)O)CO)CO)CO)CO)CO)CO)O)N)O
Color White
Quantity 200mg
Formula Weight 1134.00
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (T)
Chemical Name or Material 3A-Amino-3A-deoxy-(2AS,3AS)-beta-cyclodextrin Hydrate