Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3A-Amino-3A-deoxy-(2AS,3AS)-gamma-cyclodextrin Hydrate 94.0+%, TCI America™

Click to view available options
Quantity:
200mg
1g
189307-64-0
C48H81NO39
1296.144
UCNMOVNZUVXILW-OJHPOGNJSA-N
11535496
C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(C(C8O)O)OC9C(OC(O2)C(C9O)O)CO)CO)CO)CO)CO)CO)CO)O)N)O
189307-64-0
C48H81NO39
1296.144
UCNMOVNZUVXILW-OJHPOGNJSA-N
11535496
C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(C(C8O)O)OC9C(OC(O2)C(C9O)O)CO)CO)CO)CO)CO)CO)CO)O)N)O
Color White
Quantity 200mg
Formula Weight 1296.14
Physical Form Crystal-Powder at 20°C
Percent Purity ≥94.0% (HPLC)
Chemical Name or Material 3A-Amino-3A-deoxy-(2AS,3AS)-gamma-cyclodextrin Hydrate