Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Anatomy Posters
SDPSCI_ED

Visual aids to enhance anatomy lessons.

Supplier:  3B Scientific™ VR1171L

 View more versions of this product

Catalog No. S90754


Explore available promotions
Add to Cart

Description

Description

  • Printed on premium glossy (200g) paper
  • With two-sided lamination (125 micron) and metal eyelets
  • Dimensions: 20W × 26 in.H (50.8 × 66.0cm)

Specifications

Specifications

Hand and Wrist - Anatomy and Pathology Chart
26 in.
20 in.
Depicts the anatomy and pathology of the hand and wrist in bright colors
Science
66 cm
50.8 cm
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.