Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3B Scientific™ Anatomy Posters
Visual aids to enhance anatomy lessons.
Supplier: 3B Scientific™ VR1283L
Description
- Printed on premium glossy (200g) paper
- With two-sided lamination (125 micron) and metal eyelets
- Dimensions: 20W × 26 in.H (50.8 × 66.0cm)
Specifications
The Skin Chart | |
26 in. | |
20 in. | |
Anatomy of the Human skin, including all skin layers, as well as some common skin pathologies are displayed |
Science | |
66 cm | |
50.8 cm | |
Great addition to any doctor's office or classroom, useful tool for patient education |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction