Learn More
3B Scientific™ Deluxe Human Skeleton - includes 3B Smart Anatomy Products for Science Education
Medical-grade skeleton works for both instructional illustration and chiropractical demonstration.
Manufacturer:  3B Scientific™ A13/1
 View more versions of this product
Catalog No. S171564
Add to cart
Includes:
  • Dust cover
  • Manual
Description
Description
3B Smart Anatomy is now included with any 3B Scientific anatomy model.
Every original 3B Scientific anatomy model now includes these additional FREE features:
  • Free access to the anatomy course 3B Smart Anatomy, hosted inside the award-winning Complete Anatomy app by 3D4Medical
  • The 3B Smart Anatomy course includes 23 digital anatomy lectures, 117 different virtual anatomy models and 39 anatomy quizzes to test your knowledge
  • FREE warranty upgrade from 3 to 5 years with every product registration
TIPP: you will also receive access to a free 3-day trial to all premium features of the Complete Anatomy app when you sign up for your 3B Smart Anatomy course.
To unlock these benefits, simply scan the label located on your model and register online. All 3B Smart Anatomy features are completely free of charge.
Highly detailed, unbreakable plastic model is natural cast and presents muscle paintings, joint ligaments, numeration of bones and flexible spine with nerve endings.
  • Deluxe skull is dissectible into calvarium, base of skull and lower jaw
  • Position and spacing of teeth are reproduced from a natural denture
  • Special mounting of rib cage maintains proper positioning
  • Detachable skull, left arm and leg
  • Right side presents muscle insertions in blue and origins in red; left side of skeleton displays more than 300 nominated bones, fissures and foramina
  • M. iliocostalis and M. longissiumus are color-contrasted for clarity
  • Shoulder, elbow, hip and knee of the right side provided with life-like ligaments and movable joints
  • Flexible spine allows natural movement and features a herniated disc, protruding nerves and vertebral artery
  • Rotation and movement of skull on first and second cervical vertebrae can also be demonstrated
  • Available mounted with pelvic mount or on hanging stand
  • Height: 67 in. (170cm)
Specifications
Specifications
Education
75.8 in.
3-parts assembled skull with individually inserted teeth, detachable skull, left arm and leg, special mounting of rib cage, over 600 hand-numbered, labeled details dust cover, manual
Stable Metal Stand with 5 Casters
21.98 lb.
For both instructional illustration and chiropractical demonstration
192.5cm
Plastic
Skeleton model with muscles and ligaments
9.97kg