Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Deluxe Human Torsos Products for Science Education

28- or 24-part models are precision-crafted and hand-painted.

Manufacturer:  3B Scientific™ B30

 View more versions of this product

Catalog No. S17150A


Add to cart

Includes: Both models include:
  • Three-part removable head with partially exposed mouth and nasopharynx, half brain and arteries
  • Two-part stomach
  • Female breast plate with rib insertions
  • Right and left lungs
  • Two-part heart
  • Liver with gallbladder
  • Three-part large intestine with appendix flap
  • Small intestine
  • Front half of kidney Four-part male genital insert Three-part female genital insert with embryo
  • Instruction guide
28-part model also includes:
  • Open neck and back to study vertebrae, intervertebral discs, spinal cord, spinal nerves and vertebral arteries
  • Removable seventh thoracic vertebra
  • Eye with optic nerve
  • Four-part brain

Description

Description

  • Detailed brushwork by dedicated artisans depicts the most life-like colors of internal organs
  • Removable internal organs are meticulously molded to fit securely into their proper anatomical positions, removing unnatural gaps
  • Made of unbreakable, vinyl plastic for durable, long-lasting use
  • Male and female genital organs can be interchanged
  • Mounted on a sturdy base for support
  • Dimensions: 14.96 x 9.84 x 34.25 in. (38 x 25 x 87cm)

Specifications

Specifications

Great Human anatomy educational tool and makes learning the location of the Human organs easy
Vinyl
Deluxe dual sex torso model
7.5kg
3-parts head, 2-parts stomach, female chest wall, 2 lungs, 2-parts heart, liver with gall bladder, 4-part intestinal tract, front half of kidney, 4-part male genital insert, 3-parts female genital insert with fetus, 3B torso guide
Sturdy Base
16.53 lb.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit