Learn More
Fisherbrand™ 3D Platform Rotators Products for Science Education
Variable speed, fixed-angle 3D rotators
Catalog No. S35845
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
These fixed-angle rotators provide smooth orbital motion for mixing in culture flasks, dishes, boxes and tubes.
  • Simple speed adjustment from 10 to 30rpm
  • Gentle movement ideal for staining gels without destroying the edges or keeping fragile tissue intact during incubation
  • Large platform accommodates most commonly used vessels with non-slip mat that prevents slipping
  • Low voltage power supply provides safe cold room operation and low energy consumption
  • Operating temperature range of 4° to 40°C allows for use in cold rooms or incubators
Specifications
Specifications
10 to 30rpm
8.19 in.
20.8cm
12V
4 to 40°C
0.49kg
8.19 in.
20.8cm
0.5A
39 to 104°F