Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3H-1,2,3-Triazolo[4,5-b]pyridin-3-ol 98.0+%, TCI America™

Click to view available options
Quantity:
1g
5g
25g
39968-33-7
C5H4N4O
136.114
MFCD00210053
FPIRBHDGWMWJEP-UHFFFAOYSA-N
1-hydroxy-7-azabenzotriazole,hoat,3h-1,2,3 triazolo 4,5-b pyridin-3-ol,3-hydroxytriazolo 4,5-b pyridine,unii-tx8xyh09h0,1-hydroxy-7-azabenzotriazole hoat,hoat 1-hydroxy-7-azabenzotriazole,3h-1,2,3-triazolo 4,5-b pyridine, 3-hydroxy,tx8xyh09h0,7-aza-1-hydroxybenzotriazole
181649
3-hydroxytriazolo[4,5-b]pyridine
C1=CC2=C(N=C1)N(N=N2)O
39968-33-7
C5H4N4O
136.114
MFCD00210053
FPIRBHDGWMWJEP-UHFFFAOYSA-N
1-hydroxy-7-azabenzotriazole,hoat,3h-1,2,3 triazolo 4,5-b pyridin-3-ol,3-hydroxytriazolo 4,5-b pyridine,unii-tx8xyh09h0,1-hydroxy-7-azabenzotriazole hoat,hoat 1-hydroxy-7-azabenzotriazole,3h-1,2,3-triazolo 4,5-b pyridine, 3-hydroxy,tx8xyh09h0,7-aza-1-hydroxybenzotriazole
181649
3-hydroxytriazolo[4,5-b]pyridine
C1=CC2=C(N=C1)N(N=N2)O
Color White-Yellow
Quantity 1g
UN Number 0481
Formula Weight 136.11
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material 3H-1,2,3-Triazolo[4,5-b]pyridin-3-ol