Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3H-1,2-benzodithiol-3-one 1,1-dioxide, 98%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
66304-01-6
C7H4O3S2
200.226
MFCD00132960
JUDOLRSMWHVKGX-UHFFFAOYSA-N
3h-1,2-benzodithiol-3-one 1,1-dioxide,beaucage reagent,3h-1,2-benzodithiol-3-one-1,1-dioxide,3h-benzo c 1,2 dithiol-3-one 1,1-dioxide,3h-1,2-benzodithiol-3-one, 1,1-dioxide,1,1-dioxo-1$l^ 6 ,2-benzodithiol-3-one,3h-1,2-benzodithiol-one 1,1-dioxide,3h-1,2-benzodithiol-3-one1,1-dioxide,1??,2-benzodithiole-1,1,3-trione,acmc-1b2sk
3009847
1,1-dioxo-1$l^{6},2-benzodithiol-3-one
C1=CC=C2C(=C1)C(=O)SS2(=O)=O
66304-01-6
C7H4O3S2
200.226
MFCD00132960
JUDOLRSMWHVKGX-UHFFFAOYSA-N
3h-1,2-benzodithiol-3-one 1,1-dioxide,beaucage reagent,3h-1,2-benzodithiol-3-one-1,1-dioxide,3h-benzo c 1,2 dithiol-3-one 1,1-dioxide,3h-1,2-benzodithiol-3-one, 1,1-dioxide,1,1-dioxo-1$l^ 6 ,2-benzodithiol-3-one,3h-1,2-benzodithiol-one 1,1-dioxide,3h-1,2-benzodithiol-3-one1,1-dioxide,1??,2-benzodithiole-1,1,3-trione,acmc-1b2sk
3009847
1,1-dioxo-1$l^{6},2-benzodithiol-3-one
C1=CC=C2C(=C1)C(=O)SS2(=O)=O
Melting Point 102°C to 107°C
Packaging Glass bottle
Quantity 250mg
Color White
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 200.24
Physical Form Crystals
Percent Purity ≥97.5%
Chemical Name or Material 3H-1, 2-Benzodithiol-3-one 1, 1-dioxide
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning