Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3H-1,2-Benzodithiol-one 1,1-dioxide, 98%, Thermo Scientific Chemicals

Catalog Number aal20129md
Click to view available options
Quantity:
250 mg
5 g
66304-01-6
C7H4O3S2
200.226
MFCD00132960
JUDOLRSMWHVKGX-UHFFFAOYSA-N
3009847
1,1-dioxo-1$l^{6},2-benzodithiol-3-one
C1=CC=C2C(=C1)C(=O)SS2(=O)=O
66304-01-6
C7H4O3S2
200.226
MFCD00132960
JUDOLRSMWHVKGX-UHFFFAOYSA-N
3009847
1,1-dioxo-1$l^{6},2-benzodithiol-3-one
C1=CC=C2C(=C1)C(=O)SS2(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 102°C to 107°C
Quantity 250 mg
UN Number UN3335
Beilstein 4180332
Synonym 3h-1,2-benzodithiol-3-one 1,1-dioxide, beaucage reagent, 3h-1,2-benzodithiol-3-one-1,1-dioxide, 3h-benzo c 1,2 dithiol-3-one 1,1-dioxide, 3h-1,2-benzodithiol-3-one, 1,1-dioxide, 1,1-dioxo-1$l^ 6 ,2-benzodithiol-3-one, 3h-1,2-benzodithiol-one 1,1-dioxide, 3h-1,2-benzodithiol-3-one1,1-dioxide, 1??,2-benzodithiole-1,1,3-trione, acmc-1b2sk
Formula Weight 200.24
Percent Purity 99%
Chemical Name or Material 3H-1,2-Benzodithiol-one 1,1-dioxide
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit