Learn More
3M™ 4510 Series Disposable Protective Coveralls
Manufacturer:  3M™ 45103XL
 View more versions of this product
Catalog No. 17-623-339
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
EN 13034 Type 6, EN ISO 13982-1 Type 5, EN 1149-1:2006, ASTM D2859-96, CE
Two-way Zipper with Storm Flap
Elastic
Microporous Polyethylene
White
49 to 52 in.
124.46 to 132.08cm
194 to 200cm
Yes
54 in.
137.16cm
Anti-static
White Polypropylene/Polyethylene laminate film suit, Nylon on Polyester Braid zipper, Neoprene Rubber elastic, Polyester thread
Elastic
Bound
3X-Large
Yes
6.33 to 6.5 ft.
Paint spraying, Metal polishing, machine or vehicle maintenance (Oils and greases) and general industrial clean-up and processing, protect against light liquid splashes and hazardous dusts
35 in.
88.9cm