3M™ 4510 Series Disposable Protective Coveralls

Manufacturer: 3M™ 45104XL

 View more versions of this product

Catalog No. 17-623-340

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Certifications/Compliance EN 13034 Type 6, EN ISO 13982-1 Type 5, EN 1149-1:2006, ASTM D2859-96, CE
    Material Microporous Polyethylene
    Seam Type Bound
    Color White
    Size 4X-Large
    Coating Anti-static
    Disposable Yes
    Height (Metric) 200 to 206cm
    For Use With (Application) Paint spraying, Metal polishing, machine or vehicle maintenance (Oils and greases) and general industrial clean-up and processing, protect against light liquid splashes and hazardous dusts
    Latex Free Yes
    Closure Type Two-way Zipper with Storm Flap
    Size (English) Inseam 36 in.
    Size (Metric) Inseam 91.44cm
    Size (Metric) Waist 139.7cm
    Construction White Polypropylene/Polyethylene laminate film suit, Nylon on Polyester Braid zipper, Neoprene Rubber elastic, Polyester thread
    Ankle Style Elastic
    Size (English) Chest 52 to 55 in.
    Size (Metric) Chest 132.08 to 139.7cm
    Height (English) 6.5 to 6.75 ft.
    Size (English) Waist 55 in.
    Cuff Style Elastic

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit