Learn More
3M™ 5-Way Penetrant
Frees rusted, frozen nuts and bolts
Manufacturer:  3M™ 7000028601
Catalog No. 19-047-238
Add to cart
Description
Description
Designed to penetrate, lubricate, demoisturize, and protect delicate electrical apparatus and precision instruments. An ideal tapping lubricant for stainless steel and aluminum, it also inhibits corrosion and pitting of injection molding dyes and extension screws.
  • Provides protection against electrical failures caused by moisture and corrosion
  • Protects components such as switches, relays, batteries, motors, and generators
  • Keeps components highly efficient, even in humid environments
  • 3M Catalog# -WAYPENETRANT
Order Info
24 fl. oz. (710mL) can.
Specifications
Specifications
12 Case
Protect delicate electrical apparatus and precision instruments and an ideal tapping lubricant for stainless steel and aluminum
Safety
24 oz.
5 Way Penetrant Aerosol