3M™ 6000 Series Drop-Down Half Facepiece
Designed for workers who need to communicate frequently
Manufacturer: 3M™ 6200DD
 View more versions of this product
Catalog No. 19-041-418
  Check Availability
Add to Cart
Required Accessories : Facepiece requires compatible cartridges and/or filters. (See separate listings.)
    Specifications
    For Use With (Application) Demolition, facility maintenance, sanding
    For Use With (Equipment) Filter 2000 series, filter 7093, p100 or cartridge 6000 series,Filter 2000 series, filter 7093, p100 or cartridge 6000 series
    Product Type Drop Down Half Facepiece Respirator
    Material Thermoplastic Elastomer
    Size Medium,Medium
    Certifications NIOSH 42CFR84
    Connection Type Bayonet
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback