Learn More
3M™ 7093CB Hydrogen Fluoride Cartridge/Filter
Manufacturer:  3M™ 7000126938
Catalog No. 19-168-134
Add to cart
Description
Description
  • 3M Catalog# 7093CB
Specifications
Specifications
NIOSH
5.8cm
8.6cm
Demolition, facility maintenance, sanding
Cartridge and Filter
Filter Combo Cartridge
2.3 in.
3.4 in.
144 Case
3M™ half and full facepiece 6000, 7000, ff-400 series
Olive, Magenta