3M™ Accessories for Dual Airline Supplied Air Respirators

For use with dual airline supplied air respirator

Manufacturer: 3M™ SA1027

 View more versions of this product

Catalog No. 19-813-790

  Check Availability
Add to Cart

    Specifications

    Quantity 1 Case
    For Use With (Equipment) 3M™ low pressure dual airline supplied air respirator,3M™ low pressure dual airline supplied air respirator
    Product Type Airline Connector Valve Assembly
    Includes Dual airline connector, waist belt
    Color Gray

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit