Learn More
3M™ Adflo™ PAPR Accessory, Battery
Replacement battery for 3M™ Adflo™ Powered Air Purifying Respirator (PAPR). Environmentally-friendly NiMH battery charges in less than 3 hours. Battery can be removed or installed in turbo unit during charging.
Manufacturer:  3M™ 15109907
Catalog No. 19-808-043
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • NiMH Battery
  • Replacement Part
Specifications
Specifications
NiMH
Black
Adflo Powered Air Purifying Respirator System
Battery
OSHA, 29 CFR 1910.1200
Automotive, construction, food and beverage manufacturing, manufacturing, military maintenance, repair and operation, mining, oil and gas, transportation
Safety
15-1099-07