Learn More
3M™ Adflo™ PAPR Accessory, Breathing Tube
Manufacturer:  3M™ 15009910
Catalog No. 19-150-0595
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Adflo Powered Air Purifying Respirator System
Assembly, facility maintenance, machine operation, painting, sanding
Rubber
Breathing Tube
Gasket (15-0099-110 and Breathing tube rubber O-ring (15-0099-12)
Helmet assembly to turbo unit