3M™ Air-Mate™ Head Covers

Provide protection for a range of applications

Manufacturer: 3M™ BE-12B-50

 View more versions of this product

Catalog No. 19-813-718

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    For Use With (Equipment) 3M™ air-mate™ and breathe easy™ powered air purifying respirator system,3M™ air-mate™ and breathe easy™ powered air purifying respirator system
    Product Type Headcover
    For Use With (Application) Assembly, cleaning, electrical, facility maintenance, machine operation, painting, sanding, welding
    Material Polyethylene
    Size Regular
    Certifications NIOSH
    Color Blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit