Learn More
3M™ Air-Mate™ Vinyl Belt-Mounted High Efficiency Powered Air-Purifying Respirator System
Ideal for use in healthcare isolation wards and first receiver environments requiring easy decontamination of equipment.
Manufacturer:  3M™ AMH12U
Catalog No. 19-813-750
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product.
Includes: System includes the following components (also available separately, as noted in parentheses):
  • Air-Mate PAPR assembly 231-01-30U
  • Vinyl belt GVP-117 (Cat. No. 19-813-773 )
  • Head cover BE-12-3, Regular, White (3 pack) (Cat. No. 18-778A)
  • Smart battery charger 520-03-73 (Cat. No. 18-778A)
  • External charging assembly 520-04-24 (Cat. No 19-160-0123)
  • Breathing tube BE-224 (Cat. No. 18-039)
  • Breathing tube covers W-3228-10 (10 pack) (Cat. N0. 19-021-574)
  • Blue storage bag
Description
Description
  • Protects against airborne particulates
  • Includes three head covers: vinyl belt, breathing tube covers and external charge adaptor
  • Belt-mounted blower unit draws air through HE filter or cartridge conducts filtered air through breathing tube to head covering
  • NIOSH-approved filter offers 99.97% efficiency at 0.3µ particle size
  • Can be worn with facial hair and glasses
  • No fit testing required prior to use
  • Battery offers up to eight hours of service per charge
Specifications
Specifications
NIOSH
HE Air Filter
Air-mate™ powered air purifying respirator assembly, vinyl belt, head cover, smart battery charger, external charging assembly, breathing tube, breathing tube cover, blue storage bag
Belt
Air Purifying Respirator
Pharmaceutical, hospital and public health
Vinyl