Learn More
3M™ Airline Adapter Kits
Manufacturer:  3M™ W306025
 View more versions of this product
Catalog No. 19-021650
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Facepiece of supplied air respirator
Adapter Kit
Convert 6000din and 7000 series facepiece to airline mode
3m™ air regulating valve kit w-3061 and 3m™ supplied air hose w-9435-25