Learn More
3M™ Airstream™ Headgear-Mounted PAPR and Accessories
Provides high efficiency, integrated head, face and respiratory protection against particles without the need for a tight faceseal or fit testing
Manufacturer:  3M™ 7000126321
 View more versions of this product
Catalog No. 19-038-882
Add to cart
Includes: Higher airflow motor
Description
Description
  • Keeps faceshield fog-free
  • Constant flow of air
  • With rechargeable, NiCd battery-powered blower, HE filter and hardhat with visor
  • Up to eight hours of battery service
  • Suited for smelting, foundry and metalworking
  • 3M Catalog# AS-140-5 , 3M Catalog# AS-400LBC
Mining, oil and gas industries
Certifications
ANSI Z87.1-2003 (Eye and Face Protection) and ANSI Z89.1-2003 Type I, Class C (Industrial Head Protection)
Specifications
Specifications
Filter
HE Filter
Headgear mounted powered air purifying respirator
Safety
Helps filter contaminants before air reaches workers breathing zone
AS-140-5