Learn More
3M™ Airstream™ Headgear-Mounted PAPR and Accessories
Provides high efficiency, integrated head, face and respiratory protection against particles without the need for a tight faceseal or fit testing
Supplier: 3M™ 7000052120
Includes: Higher airflow motor
Description
- Keeps faceshield fog-free
- Constant flow of air
- With rechargeable, NiCd battery-powered blower, HE filter and hardhat with visor
- Up to eight hours of battery service
- Suited for smelting, foundry and metalworking
- 3M Catalog# AS-140-5 , 3M Catalog# AS-400LBC
Mining, oil and gas industries
Certifications
ANSI Z87.1-2003 (Eye and Face Protection) and ANSI Z89.1-2003 Type I, Class C (Industrial Head Protection)
Specifications
High Efficiency (HE) | |
Smelting, foundry and metal working, mining, oil and gas industries | |
AS-400LBC |
PAPR Hard Hat System | |
Higher airflow motor, rechargeable, nicd battery-powered blower, he filter and hardhat with visor | |
Head-mounted |