Learn More
3M™ Belt-Mounted and Vehicle-Mounted PAPR System Accessory, Cartridges
Choose from a large selection of cartridges for powered air purifying respirators used in a variety of industrial environments.
Supplier: 3M™ GVP403
Description
- Cartridge attaches to the motor blower
- Filters supply air to PAPR headgear of choice
- Products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product
- 3M Catalog# GVP-440, GVP-441
Specifications
Organic Vapor, Acid Gas | |
Yellow | |
Mining, Oil & Gas | |
GVP403 |
NIOSH | |
Cartridge | |
Belt-mounted and vehicle-mounted PAPR systems | |
6/Cs. |