3M™ Breathe-Easy™ Belt-Mounted PAPR System Breathing Tubes
Flexible, accordion-style hoses connect Turbo PAPR assembly to headgear
Manufacturer: 3M™ 526-01-13R01
 View more versions of this product
Catalog No. 19-526-013
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Alert
All safety products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product. Misuse may result in sickness or death.
    Specifications
    For Use With (Equipment) Breathe easy™ 1, 17 and 3M™ airstream™ high efficiency powered air purifying respirator system
    Product Type Breathing Tubes
    For Use With (Application) General purpose and welding headgear
    Connection Type Belt Mounted
  • Description & Specifications
Related Products
Product Content Feedback