Learn More
3M™ Breathe-Easy™ Belt-Mounted PAPR System Breathing Tubes
Flexible, accordion-style hoses connect Turbo PAPR assembly to headgear
Manufacturer:  3M™ 7000131715
 View more versions of this product
Catalog No. 19-520-032
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
All safety products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product. Misuse may result in sickness or death.
Description
Description
3M Breathe Easy Belt-Mounted Turbo PAPRs
To assemble a complete system, select from the following pieces:
  • Headgear, facepiece, head cover or hood
  • Compatible breathing tube
  • Breathe Easy belt-mounted Turbo PAPR assembly
  • Appropriate cartridge/filter pack for your application
  • Battery pack and charger
  • 3M Catalog# 520-01-00R01 , 3M Catalog# 520-03-32R01 , 3M Catalog# BE-324
Order Info
Breathing tube clamps available. (See Cat. No. 19-813-724)
Specifications
Specifications
Safety
520-03-32R01
Breathing Tube
83.82cm
Belt Mounted
Breathe easy™ 12, powered air purifying respirator system
33 in.