Learn More
3M™ Breathe Easy™ Belt-Mounted Turbo PAPR Assemblies
Choose from two styles
Supplier: 3M™ 7000002111
Description
Each assembly includes motor blower, belt, airflow indicator, and battery pack. For use with appropriate 3M filter, filter/cartridge or canister, and respirator headpiece.
3M Breathe Easy Belt-Mounted PAPRs
To assemble a complete system, select from the following pieces:
- Headgear, facepiece, head cover or hood
- Compatible breathing tube
- Breathe Easy belt-mounted Turbo PAPR assembly
- Appropriate cartridge/filter pack for your application
- Battery pack and charger
- 3M Catalog# 520-17-00
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product.
Specifications
High Efficiency (HE) | |
450-00-01r12 he (p3) filter, intrinsically safe | |
520-17-00 |
PAPR Assembly, No Headtop | |
Motor blower, belt, airflow indicator, battery pack |