Learn More
3M™ Breathe Easy™ 12 Bio Conversion Kit, Large
Easily convert your current Breathe Easy Powered Air Purifying Respirator systems that uses the BE-10BR Butyl Rubber Hood and the FR-57 cartridge for chemical protection with the BE 12 Biokit.
Manufacturer:  3M™ BE12LBIOKIT
Catalog No. 17-987-255
  Check Availability
Add to cart
Specifications
Description
Kit Components
  • BE-12L-3 Head Cover, White, Large, fabric with polyethylene coating (3 pack)
  • 520-03-32R01 Breathing Tube
  • 50-00-01 High Efficiency Filter, P3 (3 pack)
Description & Specifications
Specifications
Breathe Easy Powered Air Purifying Respirator System
Kit
Protection against biological hazards