3M™ Breathe Easy™ Filters, Cartridges, and Canisters

Manufacturer: 3M™ 4530101R06

 View more versions of this product

Catalog No. 17-987-272H

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Quantity 6 Pack
    For Use With (Equipment) 3M™ breathe easy™ powered air purifying respirator,3M™ breathe easy™ powered air purifying respirator
    Product Type Ep3 Cartridge /Filter
    For Use With (Application) For protection against sulfur dioxide, chlorine, formaldehyde, hydrogen chloride and high efficiency particulates
    Certifications NIOSH MSHA
    Cartridge Type Formaldehyde, Chlorine, Hydrogen Chloride, Sulfur Dioxide Cartridge
    Filter Type HE Filter
    Color Magenta, White

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit