3M™ Breathe Easy™ Powered Air-Purifying Respirator System with Butyl Rubber Hood
Available in NIOSH CBRN-approved configurations
Manufacturer: 3M™ RBEHUP
 View more versions of this product
Catalog No. 19-171-185
    N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
    Specifications
    Certifications NIOSH
    For Use With (Equipment) Powered air purifying respirator
    Filter Type HE Filter
    Includes Breathing tube rbe-bth and clamp, motor blower assembly with decon belt, 3m™ cartridge rbe-57, airflow indicator, choice of battery pack, butyl rubber hood
    Weight (English) 1.5 lb.
    Weight (Metric) 0.73kg
    Product Type Powered Air Purifying Respirator Conversion Kit
    For Use With (Application) For converting Breathe Easy™ (be) 10 powered air purifying respirator to niosh CBRN-approved system
    Mounting Type Belt
  • Description & Specifications
Related Products
Product Content Feedback