3M™ Canister FR-40 and Cartridge FR-57

Tested to military standards

Manufacturer: 3M™ 4530303R06

 View more versions of this product

Catalog No. 19-045-576

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Quantity 6 Pack
    For Use With (Equipment) Rrpas 6000 series tight-fitting facepiece powered air purifying respirator system,Rrpas 6000 series tight-fitting facepiece powered air purifying respirator system
    Product Type Fr-40 Canister Respirator
    Certifications NIOSH
    Filter Type High Efficiency Filter
    Color Magenta, Olive
    For Use With (Application) Protection against organic vapor, acid gas, chemical warfare agents, toxic industrial chemicals

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit