3M™ Classic Soft Ear Plugs

Manufacturer: 3M™ 3116000

 View more versions of this product

Catalog No. 19-123-295

  Check Availability
Add to Cart

    Specifications

    Product Type Classic Soft™ Earplug
    Material Polyurethane
    Shape Cylindrical
    Style Uncorded
    Noise Reduction Range (NRR) 31dB
    Certifications/Compliance CSA Class AL
    Size One size fits to all
    Color Yellow
    For Use With (Application) Plugs start out firm for easy fitting then soften from body heat for a comfortable fit

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit