Learn More
3M™ Particulate Filter P100 with Nuisance Level OV Relief
Filter provides versatile protection against many oil and non-oil based particulate contaminants.
Manufacturer:  3M™ 5057400000
 View more versions of this product
Catalog No. 19-145-450
Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Made with proprietary Advanced Electrostatic Media (AEM)
  • Particulate filters are both flame and water resistant
  • Easy to install
  • Particulate Filters 2000 Series are compatible with Half Facepieces 6000, 6500/6500QL and 7500 Series, and Full Facepieces 6000, FF-400 and 7800 Series
Specifications
Specifications
OV
White
Filter Clip
3M 6000, 6500, 7500, or FF-400
Bayonet
P
100
50574-00000