Learn More
3M™ Particulate Filter P100 with Nuisance Level OV Relief
Filter provides versatile protection against many oil and non-oil based particulate contaminants.
Specifications
Cartridge Type | OV |
---|---|
Connection Type | Bayonet |
Filter Type | P |
Filter Efficiency | 100 |
For Use With (Equipment) | 3M 6000, 6500, 7500, or FF-400 |
Catalog Number | Mfr. No. | Product Type | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Color | Price | Quantity & Availability | ||||
19-145-445
|
3M™
5057000000 |
Filter | Pink | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
19-145-450
|
3M™
5057400000 |
Filter Clip | White | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Made with proprietary Advanced Electrostatic Media (AEM)
- Particulate filters are both flame and water resistant
- Easy to install
- Particulate Filters 2000 Series are compatible with Half Facepieces 6000, 6500/6500QL and 7500 Series, and Full Facepieces 6000, FF-400 and 7800 Series
Specifications
OV | |
P | |
3M 6000, 6500, 7500, or FF-400 |
Bayonet | |
100 |