3M™ Disposable Filtering Welding and Metal Pouring Respirator, N95

For use when welding, torch cutting, soldering, brazing and working with molten metals

Manufacturer: 3M™ 8514

 View more versions of this product

Catalog No. 19-037059

  null
Add to Cart

Alert

Do not exceed maximum use concentration established by regulatory standards. If respirator becomes damaged, soiled, or breathing becomes difficult, leave the contaminated area and dispose of the respirator. Before each use of the product, please inspect for any rips, damage, tears, etc. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Certifications TC-84A-0454
    Quantity 80 Case
    Disposable Yes
    Filter Type N95 Filter
    For Use With (Application) Welding, torch cutting, soldering, brazing and working with molten metals
    Latex Free Yes
    Nosepiece Type Standard
    Nuisance Odor Relief Yes
    Resistant To Non-oil based particles
    Size Standard
    With Faceseal No
    Model 8514
    Product Type Particulate Welding Respirator
    Filter Efficiency 95% against solid and liquid aerosols that do not contain oil
    Design Type Cup
    Color Code White

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit