3M™ Disposable Filtering Welding and Metal Pouring Respirator, N95
For use when welding, torch cutting, soldering, brazing and working with molten metals
Manufacturer: 3M™ 8516
 View more versions of this product
Catalog No. 19-038-990
  Check Availability
Add to Cart
Alert
Do not exceed maximum use concentration established by regulatory standards. If respirator becomes damaged, soiled, or breathing becomes difficult, leave the contaminated area and dispose of the respirator. Before each use of the product, please inspect for any rips, damage, tears, etc. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
    Specifications
    Certifications NIOSH TC-84A-1299
    Latex Free Yes
    Product Type Cup
    Quantity 80 Case
    Disposable Yes
    Filter Type N95 Filter
    For Use With (Application) Grinding, welding, aluminum industry, glass etching, chemical processing, paper processing and brewing operation where particle and nuisance level of acid gas may be present
    Nosepiece Type M-nose clip
    Size Standard
    Weight (Metric) 0.036kg
    Weight (English) 0.08 lb.
    Color White
  • Description & Specifications
Related Products
Product Content Feedback